Learn More
Aliskiren, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 173334-57-1 |
| Molecular Formula | C30H53N3O6 |
| MDL Number | MFCD09839018 |
| Quantity | 100 mg |
| SMILES | CC(C)[C@@H](CC1=CC=C(OC)C(OCCCOC)=C1)C[C@H](N)[C@@H](O)C[C@@H](C(C)C)C(NCC(C)(C)C(N)=O)=O |
| Molecular Weight (g/mol) | 551.76 |
| Percent Purity | 98% |
| Chemical Name or Material | Aliskiren |