Learn More
Alfacalcidol, TRC
Product Code. 30133907
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133907 10 mg 10mg
18013430 1 mg 1mg
17909829 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133907 Supplier Toronto Research Chemicals Supplier No. TRCA52400010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41294-56-8
Molecular Formula C27 H44 O2
Molecular Weight (g/mol) 400.64
Synonym Alfacalcidol, (5Z,7E)-9,10-Secocholesta-5,7,10(19)-triene-1α,3β-diol, 1,3-Cyclohexanediol, 5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-, (1R,3R)-, 9,10-Secocholesta-5,7,10(19)-triene-1,3-diol, (1α,3β,5Z,7E)- (9CI), (1R,3R)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-Dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-1,3-cyclohexanediol, (1S)-Hydroxycalciol, 1-Hydroxycholecalciferol, 1-Hydroxyvitamin D3, 1α(OH)D3, 1α-Hydroxycholecalciferol, 1α-Hydroxyvitamin D3, Alfarol, Alpha D3, Alphacalcidol, Alpharol, Bondiol, EinsAlpha, Etalpha, Oxydevit, Tevabone, Un Alfa, Un Alpha, α-Calcidol
IUPAC Name (1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C27H44O2/c1-18(2)8-6-9-19(3)24-13-14-25-21(10-7-15-27(24,25)5)11-12-22-16-23(28)17-26(29)20(22)4/h11-12,18-19,23-26,28-29H,4,6-10,13-17H2,1-3,5H3/b21-11+,22-12-/t19-,23-,24-,25+,26+,27-/m1/s1
Formula Weight 400.3341
Percent Purity >95%
Chemical Name or Material Alfacalcidol
Grade HPLC