missing translation for 'onlineSavingsMsg'
Learn More

Aldolase, MP Biomedicals™

Product Code. 11420042 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 11420042

Brand: MP Biomedicals 0215985925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 9024-52-6
Quantity 25 mg
Synonym D -Fructose-1, 6-bisphosphate-D -glyceraldehyde-3-phosphate lyase, E.C. 4.1.2.13
Formula Weight ∽161, 000
Physical Form Powder
Chemical Name or Material Aldolase
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.