missing translation for 'onlineSavingsMsg'
Learn More

Albuterol Sulfate, TRC

Product Code. 30318012
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318012 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318012 Supplier Toronto Research Chemicals Supplier No. TRCA514500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Salbutamol sulfate
CAS 51022-70-9
Purity Grade Notes HPLC
Molecular Formula 2 C13 H21 N O3 . H2 O4 S
Quantity 100 mg
Synonym Salbutamol sulphate, Albuterol sulphate, 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, sulfate (2:1), 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, sulfate (2:1) (salt) (9CI), (±)-Salbutamol sulfate, Aerolin, Airet, Albuterol hemisulfate, Albuterol sulfate, Almotex, Anebron, Asmadil, Asmalin, Asmanil, Asmasal, Asmatol, Asmaven, Asmavent, Asmidon, Asmol Uni-Dose, Asthalin, Broncho-Spray, Broncovaleas, Bronter, Bugonol, Butamol, Buto-Asma, Butotal, Buventol, Cetsim, Cobutolin, Dilatamol, Ecovent, Farcolin, Grafalin, Instavent, Libretin, Loftan, Medolin, Mozal, NSC 289928, Novosalmol, Parasma, Proventil, Respax, Salbetol, Salbovent, Salbron, Salbulin, Salbumol, Salbusian, Salbutalan, Salbutamol hemisulfate, Salbutamol sulfate, Salbutan, Salbutard, Salbutine, Salbutol, Salbuven, Salbuvent, Salmaplon, Salomol, Salorain, Sch 13949W sulfate, Sedalin, Suprasma, Theosal, Tobybron, Vencronyl, Venetlin, Ventelin, Ventilan, Ventimax, Ventodiscks, Ventolin Rotacaps, Ventorain, Ventorline CR, Volma, dl-Salbutamol sulfate
InChI Formula InChI=1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4)
SMILES CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2.OS(=O)(=O)O
IUPAC Name 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
Molecular Weight (g/mol) 576.7
Formula Weight 576.2717
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.