missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Albuterol Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Salbutamol sulfate |
| CAS | 51022-70-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 2 C13 H21 N O3 . H2 O4 S |
| Quantity | 25 mg |
| Synonym | Salbutamol sulphate, Albuterol sulphate, 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, sulfate (2:1), 1,3-Benzenedimethanol, α1-[[(1,1-dimethylethyl)amino]methyl]-4-hydroxy-, sulfate (2:1) (salt) (9CI), (±)-Salbutamol sulfate, Aerolin, Airet, Albuterol hemisulfate, Albuterol sulfate, Almotex, Anebron, Asmadil, Asmalin, Asmanil, Asmasal, Asmatol, Asmaven, Asmavent, Asmidon, Asmol Uni-Dose, Asthalin, Broncho-Spray, Broncovaleas, Bronter, Bugonol, Butamol, Buto-Asma, Butotal, Buventol, Cetsim, Cobutolin, Dilatamol, Ecovent, Farcolin, Grafalin, Instavent, Libretin, Loftan, Medolin, Mozal, NSC 289928, Novosalmol, Parasma, Proventil, Respax, Salbetol, Salbovent, Salbron, Salbulin, Salbumol, Salbusian, Salbutalan, Salbutamol hemisulfate, Salbutamol sulfate, Salbutan, Salbutard, Salbutine, Salbutol, Salbuven, Salbuvent, Salmaplon, Salomol, Salorain, Sch 13949W sulfate, Sedalin, Suprasma, Theosal, Tobybron, Vencronyl, Venetlin, Ventelin, Ventilan, Ventimax, Ventodiscks, Ventolin Rotacaps, Ventorain, Ventorline CR, Volma, dl-Salbutamol sulfate |
| InChI Formula | InChI=1S/2C13H21NO3.H2O4S/c2*1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;1-5(2,3)4/h2*4-6,12,14-17H,7-8H2,1-3H3;(H2,1,2,3,4) |
| SMILES | CC(C)(C)NCC(O)c1ccc(O)c(CO)c1.CC(C)(C)NCC(O)c2ccc(O)c(CO)c2.OS(=O)(=O)O |
| IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
| Molecular Weight (g/mol) | 576.7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?