missing translation for 'onlineSavingsMsg'
Learn More

Albuterol Aldehyde Hemisulfate, TRC

Product Code. 30133992
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133992 100 mg 100mg
17944905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133992 Supplier Toronto Research Chemicals Supplier No. TRCA514495100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156339-88-7
Molecular Formula C13 H19 N O3
Molecular Weight (g/mol) 237.29
Synonym 5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxybenzaldehyde, Salbutamol Sulphate Imp. D (EP)
IUPAC Name 5-[2-(tert-butylamino)-1-hydroxyethyl]-2-hydroxybenzaldehyde
SMILES CC(C)(C)NCC(O)c1ccc(O)c(C=O)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H19NO3/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15/h4-6,8,12,14,16-17H,7H2,1-3H3
Formula Weight 237.1365
Percent Purity >95%
Chemical Name or Material 5-[(1RS)-2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxybenzaldehyde
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.