missing translation for 'onlineSavingsMsg'
Learn More

β-Alanine, Millipore™

Product Code. 10568542
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10568542 250 g 250g
10448840 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10568542 Supplier Millipore Supplier No. 1010080250
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-95-9

Specifications

Melting Point 200°C (decomposition)
MDL Number MFCD00008200
Synonym β-Alanine, Ala, 3-Aminopropionic acid, β-Alanine
Density 1.44 g/cm3 at 20°C
Molecular Weight (g/mol) 89.09
Quantity 250 g
Grade Research
Chemical Name or Material β-Alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.