Learn More
Al-23 Multibore Round Tube, O.D.: 1.2mm, Two 0.3mm Bores, Thermo Scientific Chemicals
Product Code. 11388467
Change view
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
400 mm
Unit Size:
100mm
200mm
250mm
300mm
400mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388467 250 mm 250mm
11378467 100 mm 100mm
11308477 200 mm 200mm
11398467 300 mm 300mm
15411267 400 mm 400mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11388467 Supplier Thermo Scientific Alfa Aesar Supplier No. 032565.DT
 Restricted Item
Estimated Shipment: 07-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format
Denna artikel kan inte returneras. Se returpolicy
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifikationer
Chemical Name or Material Al-23 Multibore Round Tube
Name Note Two bores
Quantity 250 mm
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.
RUO – Research Use Only
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.