Learn More
Cytiva AKTA Start Chromatography System
Product Code. 15269764 Shop All Cytiva Products
Change view
Click to view available options
Flow Rate:
0.5 to 5 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
15269764 0.5 to 5 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15269764 Supplier Cytiva Supplier No. 29022094
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
AKTA start is an easy-to-use chromatography system that automates manual purification procedures using for example HiTrap columns.
AKTA start is a preparative chromatography system for laboratory scale protein purification. AKTA start is designed as a stand-alone system, with intuitive design, simple flow path, and user-friendly interface. With AKTA start it is easy to purify a wide variety of proteins using built-in quick start methods or predefined templates, or by creating your own methods. AKTA start can be combined with the Frac30 fraction collector, the user-friendly UNICORN™ start 1.0 control software, and application-focused prepacked columns for an automated solution.
AKTA start offers the following features:
  • Compact solution for quick and reliable one-step protein purification
  • Sample injection and fraction collection options
  • Quick start methods and templates for common purification techniques
  • Intuitive touchscreen display and real-time monitoring
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 0.5 to 5 mL/min.
For Use With (Application) Using Example HiTrap Columns
Height (Metric) 360 mm
Includes Standalone system with accessory box and user documentation, Fraction collector and UNICORN start software are optional modules
Operating Pressure 0.5 Mpa
Quantity 1 Ea.
Voltage 100/120/220/240 VAC
Width (Metric) 340 mm
Type AKTA Start Chromatography System