Learn More
Cytiva Akta Short Column Holder
Product Code. 11300242 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
26 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
11300242 26 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11300242 Supplier Cytiva Supplier No. 18111317
Big Savings for Better Separation
Use promo code 27992
View all promotions
In Stock
Specifications
Type Short Column Holder
Weight (Metric) 0.06 kg
Description Short Column Holder, for columns 26 mm i.d.
Diameter (Metric) Inner 26 mm
Quantity Complete packsize
For Use With (Equipment) Classic ÄKTA