Learn More
Novus Biologicals™ AKT1/2/3 Inhibitor Peptide Set
Shop All Bio Techne Products
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
Specifications
Specifications
| Host Species | Human |
| Components | Akt (Isoforms 1,2,3) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKAVTDHPDRLWAWEKF (AKT sequence: AVTDHPDRLWAWEKF). Molecular weight: 4214. Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361 |
| For Use With (Application) | Inhibition of Akt kinase activity |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Quantity | 5 mg |
| Product Type | AKT1/2/3 Inhibitor Peptide Set |
| Molecular Weight (g/mol) | 4214 |
| Inhibitors | AKT1/2/3 |
| Form | Lyophilized |
For Research Use Only