Learn More
Syngene™ Airclean™ Process Protection Microscope Hood
Product Code. 16552972 Shop All Syngene Products
Change view
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
16552972 48 in. Each
16532972 32 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16552972 Supplier Syngene™ Supplier No. SYN48PPUVCE
  • ONLINE EXCLUSIVE PRICE
    12285.00 EUR
    10844.70 EUR / Pack of 1
    Save 1440.30 EUR
    11% Off
 Restricted Item
Estimated Shipment: 27-01-2027
Process Protection with UVTect™ Microprocessor and Microscope Sash
  • AirClean™ Systems AC600 Series Vertical Laminar Flow Workstation
  • Laminar air flow is described as the smooth, fluid, parallel flow of air in a consistent direction
  • Ideal for applications that are not hazardous to the operator but require a clean workspace
  • A compact, light-weight design allows for multiple clean zones within a laboratory
  • Two-stage filtration provides clean, Class 100 laminar flow air for process protection
  • Room air first passes through an electrostatically charged pre-filter followed by a HEPA filter resulting in ISO 5 clean air
  • Independently certified to meet or exceed applicable clean bench standards
Specifications
Product Type Laminar Flow
Depth (Metric) 61 cm
Height (Metric) 81.28 cm
Width (English) 48 in.
Width (Metric) 121.92 cm