Learn More
Asecos™ Air Purifier PURIFIAIR.488
Product Code. 17210197 Shop All asecos Products
Change view
Click to view available options
Product Type:
Air Purifier
Replacement Filter
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
17210197 Replacement Filter Each
17200197 Air Purifier Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17210197 Supplier Asecos™ Supplier No. 39323
  • ONLINE EXCLUSIVE PRICE
    207.90 EUR
    176.00 EUR / Pack of 1
    Save 31.90 EUR
    15% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Air Purifier PURIFIAIR.488 reduces indoor pollutant levels with a multi-component filter system.
  • Asecos PURIFIAIR.488 can be used to reduce pollutant levels in indoor atmospheres
    • Removes mould spores, pollen, and allergens
    • Eliminates germs and bacteria and neutralises them
    • Continuously filters out fine dust particles
    • Removes unpleasant odours and harmful gases from indoor atmospheres
    • Built-in PM2.5 sensor and colour display allow users to quickly identify and easily read off the particle concentration in the indoor atmosphere in four levels from green (low) to red (high)
  • 1 device handles a volume flow rate of up to 488 m3/h and is designed for approx. 80 m3 room volume (corresponding to a surface area of 32 m2; at a room height of 2.5 m)
    • A larger number of devices should be used in larger rooms
    • Fresh air must be supplied to the rooms via natural or technical ventilation
    • Air purifiers should not be considered a substitute for a fresh air supply
    • Efficient results over time only if the filter is replaced at regularly according to the electronic display
    • Large filter units ensure a long service life
    • Low energy requirements result in low usage costs
    • Integrated humidification for a healthy and comfortable indoor climate
Specifications
Filter Type Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter
For Use With (Equipment) AIRPURIFIAIR.488
Weight (Metric) 1.519 Kg
Height (Metric) 83 mm
Depth (Metric) 559 mm
Width (Metric) 426 mm
Product Type Replacement Filter