Learn More
Lenz Laborglasintrumente™ Air Condensers without Jacket
Description
- With NS cone and drain ring
- Made of DURAN™ tubing
Specifications
Specifications
| Includes | NS cone and drain ring |
| Length (Metric) | 1000 mm |
| Material | DURAN™ tubing |
| Quantity | 1 |
| Standard Taper Joint | NS 29/32 |