Learn More
Ailanthone, MedChemExpress
Product Code. 16461658
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16461658
Brand: Medchem Express HYN1943
MedChemExpress Ailanthone (δ13-Dehydrochaparrinone) is a potent inhibitor of both full-length androgen receptor (AR) (IC50=69 nM) and constitutively active truncated AR splice variants (AR1-651 IC50=309 nM).
Ailanthone (δ13-Dehydrochaparrinone) is a potent inhibitor of both full-length androgen receptor (AR) (IC50=69 nM) and constitutively active truncated AR splice variants (AR1-651 IC50=309 nM).
Specifications
Chemical Name or Material Ailanthone
CAS 981-15-7
Color White
Purity Grade Notes Research
Molecular Formula C20H24O7
Quantity 5 mg
Synonym δ13-Dehydrochaparrinone
Solubility Information DMSO : 50 mg/mL (132.84 mM; Need ultrasonic)
SMILES C[C@@]1([C@@H]2O)[C@]([C@@]3([C@@H]4O)O)([H])[C@@]([C@](C5)([H])C4=C)(CO3)[C@@](OC5=O)([H])C[C@@]1([H])C(C)=CC2=O
Molecular Weight (g/mol) 376.4
Formula Weight 376.4
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death