Learn More
STEMCELL Technologies AggreWell™800 24-well Plate, 5 pack
Product Code. 17168081
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 Plates
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17168081 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17168081 Supplier STEMCELL Technologies Supplier No. 34815
AggreWell™800 (5 x 24-well plate)
  • AggreWell™ plates bring an easy, standardized approach to the generation of cell aggregates, including embryoid bodies (EBs) and spheroids
  • EBs and spheroids generated using AggreWell™ plates are consistent in size and shape, and are uniform within and between experiments
  • New and improved second-generation AggreWell™ plates are compatible with a variety of cell types, including ES and iPS cells, cancer cells and more
  • Enhanced optical characteristics provide crystal clear imaging
  • Note: AggreWell™ Rinsing Solution is required for optimal EB and spheroid formation
Specifications
For Use With (Application) Differentiation; Spheroid Culture; Toxicity Assay
Quantity 5 Plates