Learn More
Giese™ Agate Mortar with Pestle
Product Code. 11705146
Change view
Click to view available options
Capacity (Metric):
6 mL
8 mL
10 mL
13 mL
15 mL
24 mL
30 mL
50 mL
80 mL
130 mL
200 mL
350 mL
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11705146 10 mL Each
10428552 8 mL Each
10768031 6 mL Each
11715146 15 mL Each
10009520 13 mL Each
11725146 24 mL Each
11735146 30 mL Each
10661025 30 mL Each
11745146 50 mL Each
11803360 80 mL Each
11813360 130 mL Each
11755146 200 mL Each
11765146 350 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11705146 Supplier Giese™ Supplier No. 600M050
  • ONLINE EXCLUSIVE PRICE
    110.00 EUR
    80.45 EUR / Pack of 1
    Save 29.55 EUR
    26% Off
View all promotions
Estimated Shipment: 23-12-2026
Grind up solid chemicals and crush solids into smaller pieces with this agate mortar and pestle. Giese™ Agate Mortar with Pestle features a smooth, dense, and hard surface.
  • Polished agate construction
  • For grinding and crushing
  • Durable
  • Available in different sizes
Specifications
Product Type Mortar and Pestle
Material Agate Stone
Capacity (Metric) 10 mL