Learn More
Thermo Scientific Chemicals Agate Mortar and Pestle, 95mm OD
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Diameter (English) Outer | 3.7 in. |
| Diameter (Metric) Outer | 95mm |
| Product Type | Mortar and Pestle |
| For Use With (Equipment) | Wig-L-Bug Mixing/Grinding Mill |