Learn More
RETSCH Agate Grinding Balls
Product Code. 10573823 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
15 mm
30 mm
40 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10573823 15 mm Each
10156451 40 mm Each
10019110 30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10573823 Supplier RETSCH Supplier No. 05.368.0111
  • ONLINE EXCLUSIVE PRICE
    49.85 EUR
    28.69 EUR / Pack of 1
    Save 21.16 EUR
    42% Off
Estimated Shipment: 16-10-2026
Specifications
Product Type Grinding Balls
Diameter (Metric) 15 mm
For Use With (Equipment) PM 100, PM 200, PM 400 Planetary Ball Mills
Material Polished Agate