Learn More
RETSCH Agate Grinding Balls
Product Code. 10156451 Shop All Retsch Products
Change view
Click to view available options
Diameter (Metric):
15 mm
30 mm
40 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10156451 40 mm Each
10019110 30 mm Each
10573823 15 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10156451 Supplier RETSCH Supplier No. 05.368.0064
Estimated Shipment: 16-10-2026
Specifications
Product Type Grinding Balls
Diameter (Metric) 40 mm
For Use With (Equipment) Planetary Ball Mill PM 400
Material Agate