Learn More
Agaricinic Acid, TRC
Product Code. 30009793
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009793 100 mg 100mg
30006755 500 mg 500mg
30010397 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009793 Supplier Toronto Research Chemicals Supplier No. TRCA425628100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 666-99-9
Molecular Formula C22 H40 O7
Molecular Weight (g/mol) 416.55
Synonym Agaric acid (6CI), 2-Hydroxy-1,2,3-nonadecanetricarboxylic acid, Agaricic acid, Agaricin, Agaricinic acid, Laricic acid, NSC 60429, NSC 65690, α-Cetylcitric acid
IUPAC Name 2-hydroxynonadecane-1,2,3-tricarboxylic acid
SMILES CCCCCCCCCCCCCCCCC(C(=O)O)C(O)(CC(=O)O)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C22H40O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-18(20(25)26)22(29,21(27)28)17-19(23)24/h18,29H,2-17H2,1H3,(H,23,24)(H,25,26)(H,27,28)
Formula Weight 416.2774
Percent Purity >95%
Chemical Name or Material Agaric acid
Grade HPLC