Learn More
Brady™ Afterglow Aluminum Anti-Slip Strips
Product Code. 11722005 Shop All Brady Products
Change view
Click to view available options
Color:
Photoluminescent
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11722005 Photoluminescent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11722005 Supplier Brady™ Supplier No. 251925
  • ONLINE EXCLUSIVE PRICE
    40.30 EUR
    37.56 EUR / Pack of 1
    Save 2.74 EUR
    6% Off
Estimated Shipment: 08-01-2027
These self-adhesive aluminium Afterglow Aluminum Anti-Slip Strips are ideal for marking the edges of stairs and doorways.
  • Available in different luminous intensities.
Specifications
Description Photoluminescent Anti-skid Strips - Aluminium High intensity
Dimensions (L x W) 0.80 m x 30 mm
For Use With (Application) Exit and directional signs
Length (English) 2.6 ft.
Length (Metric) 0.8 m
Packaging Quantity 1 Ea.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness 0.6 mm
Width (English) 1.18 in.
Width (Metric) 30 mm
Color Photoluminescent
Material Aluminum