missing translation for 'onlineSavingsMsg'
Learn More

AEBSF hydrochloride, 98.6%, For HPLC analysis, MP Biomedicals™

Product Code. 11404822 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404822 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11404822 Supplier MP Biomedicals Supplier No. 0219350350
  • ONLINE EXCLUSIVE PRICE
    58.45 EUR
    55.65 EUR / 50mg
    Save 2.80 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 30827-99-7
Color Yellow
Assay Percent Range 0.986
Molecular Formula C8H11ClFNO2S
Quantity 50 mg
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
Molecular Weight (g/mol) 239.689
PubChem CID 186136
Formula Weight 239.7
Physical Form Powder
Chemical Name or Material AEBSF hydrochloride, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.