missing translation for 'onlineSavingsMsg'
Learn More

ADX-47273, MedChemExpress

Product Code. 16475518
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475518

Brand: Medchem Express HY13058

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ADX-47273 is a potent, selective and brain-penetrant mGluR5 positive allosteric modulator (PAM), with an EC50 of 0.17 μM for potentiation of glutamate (50 nM) response. ADX-47273 has antipsychotic and procognitive activities.

ADX-47273 is a potent, selective and brain-penetrant mGluR5 positive allosteric modulator (PAM), with an EC50 of 0.17 μM for potentiation of glutamate (50 nM) response. ADX-47273 has antipsychotic and procognitive activities.

Specifications

Chemical Name or Material ADX-47273
CAS 851881-60-2
Color White
Purity Grade Notes Research
Molecular Formula C20H17F2N3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 31 mg/mL (83.93 mM)
SMILES O=C(C1=CC=C(F)C=C1)N2C[C@@H](C3=NC(C4=CC=C(F)C=C4)=NO3)CCC2
Molecular Weight (g/mol) 369.36
Formula Weight 369.36
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.