Learn More
Grant Instruments™ TX150 Series Heated Circulating Bath, Stainless Steel Tank
Product Code. 12777839
Change view
Click to view available options
Depth (Metric) Interior:
300 mm
325 mm
505 mm
690 mm
Height (Metric) Interior:
150 mm
200 mm
Width (Metric) Interior:
150 mm
300 mm
Temperature Range:
-15°C to +200°C
0°C to 200°C
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Interior Height (Metric) Interior Width (Metric) Interior Temperature Range Unit Size
12777839 690 mm 200 mm 300 mm -15°C to +200°C Each
12757839 505 mm 150 mm 300 mm 0°C to 200°C Each
12747839 325 mm 150 mm 300 mm 0°C to 200°C Each
12737839 300 mm 150 mm 150 mm 0°C to 200°C Each
12767839 505 mm 200 mm 300 mm -15°C to +200°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12777839 Supplier Grant Instruments™ Supplier No. TX150ST38
  • ONLINE EXCLUSIVE PRICE
    3415.00 EUR
    3040.20 EUR / Pack of 1
    Save 374.80 EUR
    10% Off
View all promotions
Estimated Shipment: 24-02-2027
Provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150°C. Grant Instruments™ Optima™ TX150 Series Digital Heated Circulating Bath, Stainless Steel Tank combines the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of five stainless steel (ST) tanks with different capacities.
  • Stability ±0.01ºC
  • Large, bright full colour display
  • Icon driven home screen via a dial and two function buttons
  • Programming/temperature profiling (TX150, 1 program with 30 segments)
  • Model available with/without clamp (T-Clamp)
  • Low-liquid detection (float switch)
  • 5 point user calibration
  • User adjustable over temperature dial
  • 5 point user calibration
TRUSTED_SUSTAINABILITY
Specifications
Temperature Range -15°C to +200°C
Includes Includes TX150 immersion heater/stirrer with ST38 Bath
Material Stainless Steel
Width (Metric) Interior 300 mm
Height (Metric) Interior 200 mm
Depth (Metric) Interior 690 mm