Learn More
Tocris Bioscience™ ADU-S100 disodium salt
Description
ADU-S100 disodium salt is a STING agonist. It induces CD8+ T-cell mediated antitumor immunity in a rat esophageal adenocarcinoma model. In mouse tumor models, the compound induces tumor regression and a potent antitumor immune response. ADU-S100 disodium salt induces phosphorylation of TBK1 and IRF3 in murine bone marrow macrophages.
Specifications
Specifications
| Chemical Name or Material | [P(R)]-5'-O-[(R)-Hydroxymercaptophosphinyl]-P-thioadenylyl-(2'→5')-adenosine cyclic nucleotide disodium |
| CAS | 1638750-95-4 |
| Quantity | 73.5 μg |
| Target | STING Agonists |
| Molecular Formula | C20H22N10Na2O10P2S2 |
| Purity | 95% |