missing translation for 'onlineSavingsMsg'
Learn More

ADP Ribose Sodium Salt, TRC

Product Code. 30321817
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321817 10 mg 10mg
30309131 5 mg 5mg
30316601 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30321817 Supplier Toronto Research Chemicals Supplier No. TRCA20762510MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ADP Ribose Sodium Salt
Analyte or Component Names ADP Ribose Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C15 H21 N5 O14 P2 . 2 Na
Synonym Sodium (2R,3R,4R)-1-(((((((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)(hydroxy)phosphoryl)oxy)-3,4-dihydroxy-5-oxopentan-2-olate
InChI Formula InChI=1S/C15H22N5O14P2.2Na/c16-13-9-14(18-4-17-13)20(5-19-9)15-12(26)11(25)8(33-15)3-32-36(29,30)34-35(27,28)31-2-7(23)10(24)6(22)1-21;;/h1,4-8,10-12,15,22,24-26H,2-3H2,(H,27,28)(H,29,30)(H2,16,17,18);;/q-1;2*+1/p-1/t6-,7+,8+,10-,11+,12+,15+;;/m0../s1
Solution Type Neat
SMILES [Na+].[Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)(O)OC[C@@H]([O-])[C@@H](O)[C@@H](O)C=O)[C@@H](O)[C@H]3O
IUPAC Name disodium;[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl [[(2R,3R,4R)-3,4-dihydroxy-2-oxido-5-oxopentoxy]-hydroxyphosphoryl] phosphate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 603.279
Quantity 10 mg
Formula Weight 603.036
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.