missing translation for 'onlineSavingsMsg'
Learn More

Adonitol, MP Biomedicals

Product Code. 11472910 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472910 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472910 Supplier MP Biomedicals Supplier No. 0210025205
  • ONLINE EXCLUSIVE PRICE
    56.25 EUR
    53.55 EUR / 5g
    Save 2.70 EUR
    4% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A crystalline pentose alcohol (C5H12O5) formed by the reduction of ribose which does not reduce benedict solution

Specifications

CAS 488-81-3
Density 1.53g/cm3
Molecular Formula C5H12O5
MDL Number MFCD00064291
Quantity 5 g
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
SMILES C(C(C(C(CO)O)O)O)O
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 152.146
PubChem CID 6912
Formula Weight 152.15
Chemical Name or Material Adonitol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.