Learn More
Pohli™ Adhesive Tape
Product Code. 15871418
Change view
Click to view available options
Width (Metric) Exterior:
75 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Exterior Unit Size
15871418 75 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15871418 Supplier Pohli™ Supplier No. 2403216
  • ONLINE EXCLUSIVE PRICE
    436.00 EUR
    386.20 EUR / Pack of 12
    Save 49.80 EUR
    11% Off
Estimated Shipment: 07-01-2027
Adhesive tape for Pohli SUPER FSP. This tape is obligatory for the UN approval of the SUPER FSP combination packaging being valid (6PH1/X2.2/250).
  • Adhesive tape for (UN) packaging
  • Crosswise and lengthwise glass-fiber reinforced
  • 75mm wide
  • 50 m spool
  • 500 g/piece
Specifications
Quantity 12 Rolls
Width (Metric) Exterior 75 mm
Length (Metric) Exterior 50 m
For Use With (Equipment) Hand dispenser for 75 mm tapes
Color Natural
Description For UN packaging, tear-resistant until 750 N/25mm
Material Crosswise and lengthwise glassfiber reinforced tape
This tape is obligatory for the UN approval of the SUPER FSP combination packaging being valid (6PH1/X2.2/250).