Learn More
Texwipe Adhesive Floor Mats
Product Code. 11759869 Shop All Cole Parmer Products
Change view
Click to view available options
Dimensions (L x W):
18 x 36 in.
Unit Size:
Pack of 240
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11759869 18 x 36 in. Pack of 240
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11759869 Supplier Texwipe Supplier No. 0941124
  • ONLINE EXCLUSIVE PRICE
    227.09 EUR
    217.23 EUR / Pack of 240
    Save 9.86 EUR
    4% Off
Estimated Shipment: 21-12-2026
Adhesive-coated Floor Mats capture dirt and dust so you don't track it into the cleanroom.
  • Adhesive-coated polyethylene surfaces remove dirt from shoes and cart wheels.
  • When the top layer is dirty, simply peel off to expose a fresh surface.
  • Clear on White.
  • 60-layer mats.
Specifications
Dimensions (L x W) 18 x 36 in.
Quantity 4/Cs.
Color White
Length (English) 18 in.
Length (Metric) 46 cm
Material Polyethylene
Type Adhesive Floor Mat
Width (English) 36 in.
Width (Metric) 91 cm