Learn More
Adenosine, 99.8%, MP Biomedicals™
Product Code. 11398519 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398519 25 g 25g
11388519 5 g 5g
11308529 100 g 100g
11318529 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11398519 Supplier MP Biomedicals Supplier No. 0210019925
  • ONLINE EXCLUSIVE PRICE
    104.00 EUR
    98.70 EUR / 25g
    Save 5.30 EUR
    5% Off
 Restricted Item
Estimated Shipment: 18-12-2026
A nucleotide resulting from the union of one molecule of sugar -- d-ribose -- with a base, l-adenine
Chemical Identifiers
CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Specifications
Color White
Density 2.09g/cm3
Assay Percent Range 99.80%
Quantity 25 g
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Formula Weight 267.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine