Learn More
Adenosine 5’-Monophosphoramidate Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Adenosine 5'-Monophosphoramidate Sodium Salt |
| CAS | 102029-68-5 |
| Molecular Formula | C10 H14 N6 O6 P . Na |
| Quantity | 10 mg |
| Synonym | Adenosine 5'-(Hydrogen Phosphoramidate) Monosodium Salt |
| InChI Formula | InChI=1S/C10H15N6O6P.Na/c11-8-5-9(14-2-13-8)16(3-15-5)10-7(18)6(17)4(22-10)1-21-23(12,19)20;/h2-4,6-7,10,17-18H,1H2,(H2,11,13,14)(H3,12,19,20);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1 |
| SMILES | [Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)(N)[O-])[C@@H](O)[C@H]3O |
| IUPAC Name | sodium;amino-[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy]phosphinate |
| Molecular Weight (g/mol) | 368.22 |
| Formula Weight | 368.061 |