Learn More
Adenosine 5'-Monophosphomorpholidate 4-Morpholine-N,N'-dicyclohexylcarboxamidine salt, TRC
Specifications
Specifications
| Chemical Name or Material | Adenosine 5'-Monophosphomorpholidate 4-Morpholine-N,N'-dicyclohexylcarboxamidine salt |
| CAS | 24558-92-7 |
| Molecular Formula | C17 H32 N3 O . C14 H20 N6 O7 P |
| Quantity | 250 mg |
| Synonym | Adenosine 5’-Monophosphomorpholidate 4-Morpholine-N,N’-Dicyclohexylcarboxamidine Salt |
| InChI Formula | InChI=1S/C17H31N3O.C14H21N6O7P/c1-3-7-15(8-4-1)18-17(20-11-13-21-14-12-20)19-16-9-5-2-6-10-16;15-12-9-13(17-6-16-12)20(7-18-9)14-11(22)10(21)8(27-14)5-26-28(23,24)19-1-3-25-4-2-19/h15-16H,1-14H2,(H,18,19);6-8,10-11,14,21-22H,1-5H2,(H,23,24)(H2,15,16,17)/t;8-,10-,11-,14-/m.1/s1 |
| SMILES | Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)([O-])N4CCOCC4)[C@@H](O)[C@H]3O.C5CCC(CC5)[NH2+]C(=NC6CCCCC6)N7CCOCC7 |
| IUPAC Name | [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-morpholino-phosphinate;cyclohexyl-(N-cyclohexyl-C-morpholino-carbonimidoyl)ammonium |
| Molecular Weight (g/mol) | 709.77 |
| Formula Weight | 709.3676 |