missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Monophosphate Disodium Salt Hydrate, ≥99%, MP Biomedicals™

Product Code. 11473081 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473081 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473081 Supplier MP Biomedicals Supplier No. 0215026425
  • ONLINE EXCLUSIVE PRICE
    248.00 EUR
    236.25 EUR / 25g
    Save 11.75 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Source: yeast

Specifications

CAS 4578-31-8
Color White
Assay Percent Range ≥99%
Molecular Formula C10H12N5Na2O7P
MDL Number MFCD00065023
Quantity 25 g
Synonym adenosine 5'-monophosphate disodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate, disodium amp, amp disodium salt, disodium 5'-amp, adenosine phosphate disodium, 5'-amp disodium salt, 5'-adenylic acid, disodium salt, unii-t1wz11dsrn, adenylic acid disodium salt
InChI Key QGXLVXZRPRRCRP-IDIVVRGQSA-L
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
IUPAC Name disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol) 391.19
PubChem CID 20712
ChEBI CHEBI:81690
Formula Weight 391.2
Percent Purity ≥99%
Grade Not Available
Physical Form Powder
Chemical Name or Material Adenosine-5'-Monophosphate Disodium Salt Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.