Learn More
Tocris Bioscience™ Adenosine 5'-(a,b-methylene)diphosphate sodium salt
Ecto-5'-nucleotidase (CD73) inhibitor
Brand:  Tocris Bioscience™ 3633/10
Encompass
Product Code. 15786156
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Adenosine 5'-(α,β-methylene)diphosphate sodium salt is an Ecto-5'-nucleotidase/CD73 inhibitor. Blocks ecto-5'-nucleotidase-mediated adenosine production by preventing the conversion of AMP to adenosine. Reverses the doxorubicin-resistance in ecto-5'-nucleotidase-positive MDR cells (MCF7/A6 and L1210/A). Also increases rhodamine uptake and inhibits rhodamine efflux from ecto-5'-nucleotidase-positive MDR cells. Vasoconstrictor.
Specifications
Specifications
>98%
AMPCP, APCP
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions