Learn More
Tocris Bioscience™ Adenosine 5'-(a,b-methylene)diphosphate sodium salt
Ecto-5'-nucleotidase (CD73) inhibitor
Brand: Tocris Bioscience™ 3633/10
This item is not returnable.
View return policy
Description
Adenosine 5'-(α,β-methylene)diphosphate sodium salt is an Ecto-5'-nucleotidase/CD73 inhibitor. Blocks ecto-5'-nucleotidase-mediated adenosine production by preventing the conversion of AMP to adenosine. Reverses the doxorubicin-resistance in ecto-5'-nucleotidase-positive MDR cells (MCF7/A6 and L1210/A). Also increases rhodamine uptake and inhibits rhodamine efflux from ecto-5'-nucleotidase-positive MDR cells. Vasoconstrictor.Specifications
| >98% | |
| AMPCP, APCP | |
| 10 mg |
Product Title
Spot an opportunity for improvement?
For Research Use Only