Learn More
Adenosine-2-d1, CDN
Specifications
Specifications
| Chemical Name or Material | Adenosine-2-D |
| CAS | 109923-50-4 |
| Molecular Formula | C10 D H12 N5 O4 |
| Quantity | 0.01 g |
| Synonym | Adenosine-2-d (9 CI, ACI), Adenosine-2-D, 9-β-D-Ribofuranosyl-9 H-purin-2 d-6-amine, 9-β-D-Ribofuranosyl-9 H-purin-6-amine-2 d, Adenosine-2-d1 |
| InChI Formula | InChI=1 S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18 H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1/i2D |
| SMILES | [2 H]c1nc(N)c2ncn([C@@H]3 O[C@H](CO)[C@@H](O)[C@H]3 O)c2n1 |
| IUPAC Name | (2 R,3 R,4 S,5 R)-2-(6-amino-2-deuteriopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| Molecular Weight (g/mol) | 268.2475 |
| Formula Weight | 268.103 g/mol |
| Show More |