Learn More
Heidolph™ Multi-tier Attachment
Product Code. 10345632 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
25 to 250 mL Erlenmeyer Flasks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10345632 25 to 250 mL Erlenmeyer Flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10345632 Supplier Heidolph™ Supplier No. 5496200000
Estimated Shipment: 07-01-2027
For use with Unimax 2010, Promax 2020 and Polymax 2040
Includes perforated platform 2000 and 4 supporting rods
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 0 to 1100 rpm
Holds 25 to 250 mL Erlenmeyer Flasks
Product Type Support Floor