Learn More
Memmert™ Additional Stainless Steel Grid for UN55, Electropolished
Product Code. 17560286 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN55 Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17560286 UN55 Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17560286 Supplier Memmert™ Supplier No. UN55E20164
 Restricted Item
Estimated Shipment: 13-11-2026
Additional grid features a sturdy stainless steel construction.
  • Stainless Steel
  • Electropoloished
Specifications
For Use With (Equipment) UN55 Oven
Type Grid
Material Stainless Steel