Learn More
Thermo Scientific™ Adapters for TX-150 Swinging Bucket Rotors
Product Code. 15105253 Shop All Thermo Scientific Products
Change view
Click to view available options
Tube Type:
100 mL Round Bottom Open-Top Tube
11 mL IVF Tube
13 mL Urine Tube
25 mL Universal Flat/Skirted Tube
3/5 mL Blood Collection Tube
5/7 mL Blood Collection Tube
50 mL Conical or Skirted Tube
Microtube
Unit Size:
Each
Pack of 4
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Type Unit Size
15105253 5/7 mL Blood Collection Tube Pack of 4
15145263 25 mL Universal Flat/Skirted Tube Pack of 4
15165243 3/5 mL Blood Collection Tube Each
15183263 11 mL IVF Tube Pack of 4
15135243 50 mL Conical or Skirted Tube Pack of 4
15195243 100 mL Round Bottom Open-Top Tube Pack of 4
15135263 Microtube Pack of 4
15551510 13 mL Urine Tube Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15105253 Supplier Thermo Scientific™ Supplier No. 75005739
  • ONLINE EXCLUSIVE PRICE
    432.00 EUR
    346.50 EUR / Pack of 4
    Save 85.50 EUR
    19% Off
Estimated Shipment: 03-12-2026
Assorted tube adapters
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type 5/7 mL Blood Collection Tube
Capacity 5/7 mL
Max. Tube Diameter 13 mm
Max. Tube Length 116 mm
For Use With (Equipment) 50mL Conical Buckets for TX-150 rotor