Learn More
Eppendorf™ Adapter for Conical Tubes
Product Code. 10616640 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Rotor A-4-81 and Rotor FA-45-6-30
Rotor FA-45-6-30
Unit Size:
Pack of 2
Pack of 7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10616640 Rotor A-4-81 and Rotor FA-45-6-30 Pack of 7
10725642 Rotor FA-45-6-30 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10616640 Supplier Eppendorf™ Supplier No. 5820718005
  • ONLINE EXCLUSIVE PRICE
    275.00 EUR
    255.00 EUR / Pack of 7
    Save 20.00 EUR
    7% Off
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Designed for use with a variety of products to expand liquid handling capabilities in the lab
  • Available in packs of two or seven
  • For use with Eppendorf Rotors
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Conical
Capacity 15 mL
For Use With (Equipment) Rotor A-4-81 and Rotor FA-45-6-30
Color Orange
No. per Pack Set of 7