missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-d16, TRC

Product Code. 30179384
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179384 50 mg 50mg
30178930 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179384 Supplier Toronto Research Chemicals Supplier No. TRCA20780250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30470-60-1
Molecular Formula C10 D16
Molecular Weight (g/mol) 152.333
Synonym Tricyclo[3.3.1.13,7]decane-1,2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-d16 (ACI), Adamantane-d16 (8CI), Tricyclo[3.3.1.13,7]decane-d16 (9CI), 1,2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-Hexadecadeuterioadamantane, Perdeuteroadamantane, Adamantane-D16, Tricyclo[3.3.1.13,·]decane-d16
IUPAC Name 1,2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-hexadecadeuterioadamantane
SMILES [2H]C1([2H])C2([2H])C([2H])([2H])C3([2H])C([2H])([2H])C([2H])(C([2H])([2H])C1([2H])C3([2H])[2H])C2([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H16/c1-7-2-9-4-8(1)5-10(3-7)6-9/h7-10H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D,10D
Formula Weight 152.226
Chemical Name or Material Adamantane-d16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.