missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carbonitrile, 97%

Product Code. 11486917
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486917 5 g 5g
11476917 1 g 1g
11496917 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486917 Supplier Thermo Scientific Alfa Aesar Supplier No. A14352.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 23074-42-2 | C11H15N | 161.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23074-42-2
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00074731
InChI Key FQFZASRJFRAEIH-UHFFFAOYSA-N
Synonym 1-adamantanecarbonitrile, 1-cyanoadamantane, 1-adamantyl cyanide, tricyclo 3.3.1.13,7 decane-1-carbonitrile, tricyclo 3.3.1.1∼3,7∼ decane-1-carbonitrile, adamantanecarbonitrile, 3r,5s,7s-adamantane-1-carbonitrile, 1-adamantylnitrile, pubchem8757
PubChem CID 90878
IUPAC Name adamantane-1-carbonitrile
SMILES C1C2CC3CC1CC(C2)(C3)C#N

Specifications

Melting Point 193°C to 196°C
Quantity 5 g
UN Number UN3439
Formula Weight 161.25
Percent Purity 97%
Chemical Name or Material Adamantane-1-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.