missing translation for 'onlineSavingsMsg'
Learn More

AD80, MedChemExpress

Product Code. 16473658
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473658 10 mg 10mg
16446397 5 mg 5mg
16432816 25 mg 25mg
16430959 1 mg 1mg
16424746 100 mg 100mg
16410057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16473658 Supplier Medchem Express Supplier No. HY101963

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AD80, a multikinase inhibitor, inhibits RET, RAF,SRCand S6K, with greatly reduced mTOR activity.

AD80, a multikinase inhibitor, inhibits RET, RAF,SRCand S6K, with greatly reduced mTOR activity.

Specifications

Chemical Name or Material AD80
CAS 1384071-99-1
Color White
Purity Grade Notes Research
Molecular Formula C22H19F4N7O
Quantity 10 mg
Solubility Information DMSO : ≥ 150 mg/mL (316.84 mM)
SMILES O=C(NC1=CC(C(F)(F)F)=CC=C1F)NC2=CC=C(C3=NN(C(C)C)C4=NC=NC(N)=C43)C=C2
Molecular Weight (g/mol) 473.43
Formula Weight 473.43
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.