Learn More
Activator Reagent,Solstice
Product Code. 16825431
Change view
Click to view available options
Quantity:
4 x 2.5 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16825431 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16825431 Supplier Burdick & Jackson BioSyn Supplier No. BR7214X2.5L
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile
Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis
  • BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
CAS 89797-68-2, 75-05-8
CAS Max % 4.00%, 96.00%
Quantity 4 x 2.5 L
Flash Point 7°C
Packaging Glass Bottle
Physical Form Liquid
Purity Grade Notes Activator reagents
Synonym 5-(Ethylthio)-1H-Tetrazole, 5-(Ethylthio)Tetrazole Solution
Boiling Point 82°C.The physical data is that of the main component.