Learn More
Activator Reagent 0.5 M ETT in Acetonitrile, BioSyn™, Honeywell Burdick & Jackson™
BIOPHARMA
Product Code. 16845431
Change view
Click to view available options
Activator Reagent 0.5 M ETT in Acetonitrile, 4 x 2500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16845431 Activator Reagent 0.5 M ETT in Acetonitrile, 4 x 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16845431 Supplier Burdick & Jackson BioSyn Supplier No. BR7254X2.5GL
  • ONLINE EXCLUSIVE PRICE
    14425.00 EUR
    14004.41 EUR / Pack of 4
    Save 420.59 EUR
    2% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
BioSyn™ Honeywell Burdick & Jackson™ reagent solvents for DNA/RNA synthesis
  • Specifically processed and purified to exacting specifications, ensure outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of application
  • All solvents and reagents are filtered (0.2 micron for oxidation reagents and 0.1 micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Activator reagent
pH 2.5
Melting Point −46°C
CAS 75-05-8
Color Colorless
CAS Max % 0.92
Quantity 4 x 2.5 L
Packaging GL45 Glass Bottle
Physical Form Clear liquid
Product Line BioSyn
Boiling Point 8.58% (V)
Show More Show Less
Boiling and melting point temperatures are based off the physical main component.