Learn More
Activator Reagent 0.25 M ETT in Acetonitrile, BioSyn™, Honeywell Burdick & Jackson™
BIOPHARMA
Product Code. 16815431
Change view
Click to view available options
Activator Reagent 0.25 M ETT in Acetonitrile, 2500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16815431 Activator Reagent 0.25 M ETT in Acetonitrile, 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16815431 Supplier Burdick & Jackson BioSyn Supplier No. BR7212.5L
  • ONLINE EXCLUSIVE PRICE
    282.00 EUR
    268.95 EUR / Pack of 1
    Save 13.05 EUR
    4% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
BioSyn™ Honeywell Burdick & Jackson™ reagent solvents for DNA/RNA synthesis
  • Specifically processed and purified to exacting specifications, ensure outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of application
  • All solvents and reagents are filtered (0.2 micron for oxidation reagents and 0.1 micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Activator reagent
pH 2.6
Melting Point −46°C
CAS 89797-68-2
Color Colorless
CAS Max % 0.04
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Product Line BioSyn
Boiling Point 82°C
Show More Show Less
Boiling and melting point temperatures are based off the physical main component.