Learn More
Actinomycin D, >99%, MP Biomedicals™
Product Code. 11497571 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497571 10 mg 10mg
11477571 1 mg 1mg
11417581 50 mg 50mg
11427581 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11497571 Supplier MP Biomedicals Supplier No. 0210465810
  • ONLINE EXCLUSIVE PRICE
    348.00 EUR
    330.75 EUR / 10mg
    Save 17.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Chemical Identifiers
CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
Synonym Dactinomycin, Actinomycin-[thr-val-pro-sar-meval]
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
Specifica
Type Actinomycin D
Physical Form Powder
Quantity 10 mg
Assay Percent Range >99%
Formula Weight 1255.5
Grade Not Available
Chemical Name or Material Actinomycin D
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.