Learn More
Acryloylglycine, TRC
Specifications
Specifications
| Chemical Name or Material | Acryloylglycine |
| CAS | 24599-25-5 |
| Molecular Formula | C5 H7 N O3 |
| Quantity | 1 g |
| Synonym | N-(1-Oxo-2-propen-1-yl)glycine, N-(1-Oxo-2-propenyl)glycine, Acryloylaminoacetic Acid, N-(Carboxymethyl)acrylamide, N-Acryloylglycine, NSC 288735 |
| InChI Formula | InChI=1S/C5H7NO3/c1-2-4(7)6-3-5(8)9/h2H,1,3H2,(H,6,7)(H,8,9) |
| SMILES | OC(=O)CNC(=O)C=C |
| IUPAC Name | 2-(prop-2-enoylamino)acetic acid |
| Molecular Weight (g/mol) | 129.11 |
| Formula Weight | 129.0426 |