missing translation for 'onlineSavingsMsg'
Learn More

Acrylic Acid, Sigma-Aldrich™

Product Code. 15848180
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15848180 1 L 1 L
15836230 100 mL 100mL
15838180 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15848180 Supplier Sigma-Aldrich Supplier No. 8001811000
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized with hydroquinone monomethyl ether for synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-10-7
Grade Research
Molecular Formula CH2=CHCOOH
Molecular Weight (g/mol) 72.06

Specifications

Melting Point 13°C
Density 1.05 g/cm3 at 20°C
Boiling Point 141°C/1013 hPa
MDL Number MFCD00004367
Quantity 1 L
Synonym Acrylic acid, 2-Propenoic acid, Acroleic acid, Acrylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.