Learn More
Cytiva AcroPak™ 400 Capsules with Supor™ EKV Membrane
Click to view available options
Outlet Style:
1 to 1.5 in. Sanitary Flange
1/2 in. Hose Barb Inlet with Filling Bell on Outlet
Unit Size:
Each
Description
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Specifications
| Material (Membrane) | Hydrophilic PES |
| Pore Size | 0.2 μm |
| Sterility | Sterile |
| Filtration Area | 375 cm2 |
| Outlet Style | 1 to 1.5 in. Sanitary Flange |
| Quantity | 1/Ea. |